Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1993128 |
BC003779.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1993128 |
U19859.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1993128 |
NM_018855.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1993128 |
AY692443.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836943 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
AK134274.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
AK138442.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
AK146469.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
BC011321.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
BC024730.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
BC024981.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
BC033398.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807992 |
NM_153065.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
197546 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
314981 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
585853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
118360 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697661 |
AK004127.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697661 |
NM_133975.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
755904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84193 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174010 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105876 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110903 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1993128 |
BC003779.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1993128 |
U19859.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1993128 |
NM_018855.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1993128 |
AY692443.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426943 |
BC003779.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426943 |
U19859.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426943 |
NM_018855.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426943 |
AY692443.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240314 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240320 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240311 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240326 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240316 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240318 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240328 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240322 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240310 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240319 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26240323 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105876 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110903 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01850741 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06672538 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P426943 |
BC003779.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P426943 |
U19859.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P426943 |
NM_018855.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P426943 |
AY692443.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114466 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115582 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105876 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110903 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07331129 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07331118 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07331120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240334 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240398 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240298 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26240258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06760176 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240302 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240326 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240323 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06901561 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240311 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06704658 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240343 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240316 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240342 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240307 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240321 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240300 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240303 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240319 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240314 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06881781 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240305 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240320 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240333 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240345 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240318 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240329 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240336 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26240353 |
- |
- |