Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019753.4 | NP_062727.1 | 5 - 6 | 567 | 83 | |
GenBank mRNA | AF177669.1 | - | 5 - 6 | 463 | 83 | |
AK089791.1 | - | 5 - 6 | 534 | 83 | ||
AK155633.1 | - | 5 - 6 | 568 | 83 | ||
AK171733.1 | - | 5 - 6 | 490 | 83 | ||
BC042891.1 | - | 5 - 6 | 485 | 4208360 | 83 | |
D87912.1 | - | 5 - 6 | 512 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |