Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019861.1 | NP_063914.1 | 4 - 5 | 610 | 83 | |
GenBank mRNA | AF136280.1 | - | 4 - 5 | 511 | 83 | |
AF197480.1 | - | 4 - 5 | 555 | 83 | ||
AJ131851.1 | - | 4 - 5 | 487 | 83 | ||
AK017474.1 | - | 4 - 5 | 496 | 83 | ||
AK075862.1 | - | 4 - 5 | 503 | 83 | ||
AK165416.1 | - | 3 - 4 | 670 | 83 | ||
BC004054.1 | - | 1 - 2 | 81 | 83 | ||
BC051131.1 | - | 3 - 4 | 729 | 83 | ||
BC058758.1 | - | 4 - 5 | 501 | 6492834 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |