Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020284.1 | NP_064680.1 | 1 - 2 | 57 | 73 | |
GenBank mRNA | AF245399.1 | - | 1 - 2 | 57 | 73 | |
AK005429.1 | - | 1 - 2 | 77 | 73 | ||
AK014432.1 | - | 1 - 2 | 73 | 73 | ||
AY943668.1 | - | 1 - 2 | 126 | 73 | ||
BC100338.1 | - | 1 - 2 | 52 | 30712889 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |