Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140734.1 | - | 6 - 7 | 674 | 80 | |
AK220466.1 | - | 6 - 7 | 683 | 80 | ||
BC021396.1 | - | 5 - 6 | 713 | 80 | ||
BC070412.1 | - | 7 - 8 | 848 | 5705112 | 80 | |
U96963.1 | - | 5 - 6 | 686 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |