Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008052.3 | NP_032078.2 | 5 - 6 | 2035 | 65 | |
GenBank mRNA | AB015422.1 | - | 4 - 5 | 1638 | 65 | |
AK088630.1 | - | 5 - 6 | 1991 | 65 | ||
AK169455.1 | - | 5 - 6 | 2036 | 65 | ||
AK220514.1 | - | 4 - 5 | 1616 | 65 | ||
BC053055.1 | - | 5 - 6 | 1181 | 5685525 | 65 | |
BC065075.1 | - | 6 - 7 | 1817 | 65 | ||
U38252.1 | - | 3 - 4 | 1220 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |