Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008090.5 | NP_032116.4 | 4 - 5 | 1280 | 57 | |
GenBank mRNA | AB000096.2 | - | 4 - 5 | 1172 | 57 | |
AK004551.1 | - | 4 - 5 | 1277 | 57 | ||
AK004675.1 | - | 4 - 5 | 1609 | 57 | ||
AK154792.1 | - | 4 - 5 | 1278 | 57 | ||
AK154981.1 | - | 4 - 5 | 1281 | 57 | ||
BC107009.2 | - | 4 - 5 | 1184 | 40047573 | 57 | |
BC107010.1 | - | 3 - 4 | 957 | 40047574 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |