Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293690.1 | NP_001280619.1 | 4 - 5 | 600 | 83 | |
NM_008324.2 | NP_032350.1 | 4 - 5 | 447 | 83 | ||
GenBank mRNA | AK033783.1 | - | 4 - 5 | 601 | 83 | |
BC049931.1 | - | 4 - 5 | 438 | 5387937 | 83 | |
M69109.1 | - | 4 - 5 | 504 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |