Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008479.2 | NP_032505.1 | 4 - 5 | 1118 | 74 | |
GenBank mRNA | BC120591.1 | - | 4 - 5 | 1050 | 40129514 | 74 |
X98113.1 | - | 4 - 5 | 1120 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |