Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009441.2 | NP_033467.2 | 37 - 38 | 4914 | 119 | |
GenBank mRNA | AB008516.1 | - | 36 - 37 | 4791 | 119 | |
AK162004.1 | - | 2 - 3 | 219 | 119 | ||
AK164106.1 | - | 5 - 6 | 515 | 119 | ||
BC145489.1 | - | 32 - 33 | 4434 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |