Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009515.2 | NP_033541.1 | 1 - 2 | 201 | 80 | |
GenBank mRNA | BY232461.1 | - | 1 - 2 | 202 | 80 | |
U29673.1 | - | 1 - 2 | 162 | 80 | ||
U42471.1 | - | 1 - 2 | 162 | 80 | ||
U54788.1 | - | 1 - 2 | 162 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |