Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gypa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010369.3 | NP_034499.3 | 3 - 4 | 206 | 65 | |
GenBank mRNA | AK145998.1 | - | 3 - 4 | 207 | 65 | |
AK157851.1 | - | 3 - 4 | 207 | 65 | ||
AK162614.1 | - | 3 - 4 | 191 | 65 | ||
BC148192.1 | - | 3 - 4 | 470 | 40045496 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |