Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 16 - 17 | 1894 | 106 | |
GenBank mRNA | AK144304.1 | - | 17 - 18 | 2053 | 106 | |
U92949.1 | - | 16 - 17 | 1884 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |