Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011932.2 | NP_036062.1 | 4 - 5 | 622 | 81 | |
GenBank mRNA | AF163255.1 | - | 4 - 5 | 576 | 81 | |
AF186023.1 | - | 4 - 5 | 597 | 81 | ||
AK076886.1 | - | 2 - 3 | 175 | 81 | ||
AK145745.1 | - | 4 - 5 | 588 | 81 | ||
AK146180.1 | - | 4 - 5 | 591 | 81 | ||
AK169584.1 | - | 3 - 4 | 487 | 81 | ||
BC014759.1 | - | 4 - 5 | 4486834 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |