Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011994.2 | NP_036124.2 | 5 - 6 | 1704 | 65 | |
GenBank mRNA | AK051445.1 | - | 5 - 6 | 1678 | 65 | |
AK082588.1 | - | 5 - 6 | 1676 | 65 | ||
AK087653.1 | - | 5 - 6 | 1323 | 65 | ||
AK161027.1 | - | 5 - 6 | 1365 | 65 | ||
BC019187.1 | - | 5 - 6 | 1648 | 5027149 | 65 | |
Z48670.1 | - | 5 - 6 | 1708 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |