Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013711.3 | NP_038739.2 | 5 - 6 | 489 | 82 | |
GenBank mRNA | AB027566.1 | - | 5 - 6 | 489 | 82 | |
AF136399.1 | - | 5 - 6 | 492 | 82 | ||
AF171053.1 | - | 5 - 6 | 455 | 82 | ||
AF412308.1 | - | 5 - 6 | 491 | 82 | ||
AK075828.1 | - | 5 - 6 | 467 | 82 | ||
BC013688.1 | - | 5 - 6 | 469 | 3584154 | 82 | |
BC029344.1 | - | 5 - 6 | 469 | 82 | ||
BC052157.1 | - | 5 - 6 | 468 | 6506286 | 82 | |
BC052758.1 | - | 5 - 6 | 468 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |