Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016692.3 | NP_057901.2 | 13 - 14 | 2076 | 69 | |
GenBank mRNA | AB100432.1 | - | 12 - 13 | 1870 | 69 | |
AB100433.1 | - | 13 - 14 | 1882 | 69 | ||
AF117610.1 | - | 13 - 14 | 2012 | 69 | ||
AK034764.1 | - | 12 - 13 | 2336 | 69 | ||
AK081841.1 | - | 13 - 14 | 2010 | 69 | ||
AK088627.1 | - | 13 - 14 | 2033 | 69 | ||
BC052414.1 | - | 13 - 14 | 2009 | 6401188 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |