Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038710.1 | NP_001033799.1 | 2 - 3 | 565 | 111 | |
NM_016711.3 | NP_057920.2 | 2 - 3 | 501 | 111 | ||
GenBank mRNA | AF177170.1 | - | 2 - 3 | 450 | 111 | |
AF237629.1 | - | 3 - 4 | 409 | 111 | ||
AK015263.1 | - | 2 - 3 | 546 | 111 | ||
AK017072.1 | - | 2 - 3 | 546 | 111 | ||
AK076562.1 | - | 2 - 3 | 515 | 111 | ||
AK079077.1 | - | 2 - 3 | 345 | 111 | ||
AK147581.1 | - | 2 - 3 | 497 | 111 | ||
AK164178.1 | - | 2 - 3 | 347 | 111 | ||
BC052227.1 | - | 2 - 3 | 565 | 111 | ||
BC061124.1 | - | 2 - 3 | 331 | 6531518 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |