Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018695.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018695.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018695.mm.1 |
TC1500002270.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018699.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018699.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018699.mm.1 |
TC1500002270.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018698.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018698.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018698.mm.1 |
TC1500002270.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018697.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018697.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018697.mm.1 |
TC1500002270.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018696.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018696.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018696.mm.1 |
TC1500002270.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018694.mm.1 |
NM_019956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018694.mm.1 |
NM_019956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018694.mm.1 |
TC1500002270.mm.1 |
MTA TC |