Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB162929.1 | - | 6 - 7 | 812 | 102 | |
AF190699.1 | - | 6 - 7 | 884 | 102 | ||
AF190991.1 | - | 6 - 7 | 884 | 102 | ||
AK015486.1 | - | 5 - 6 | 535 | 102 | ||
AK018148.1 | - | 6 - 7 | 876 | 102 | ||
AK031550.1 | - | 6 - 7 | 912 | 102 | ||
AK042758.1 | - | 6 - 7 | 665 | 102 | ||
AK043998.1 | - | 6 - 7 | 932 | 102 | ||
AK048413.1 | - | 6 - 7 | 883 | 102 | ||
AK049903.1 | - | 5 - 6 | 618 | 102 | ||
AK081981.1 | - | 6 - 7 | 886 | 102 | ||
AK083126.1 | - | 5 - 6 | 1009 | 102 | ||
BC046310.1 | - | 6 - 7 | 870 | 6307420 | 102 | |
BC065694.1 | - | 6 - 7 | 876 | 6813365 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |