Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 5 - 6 | 485 | 61 | |
GenBank mRNA | AF211864.1 | - | 5 - 6 | 485 | 61 | |
AK089167.1 | - | 5 - 6 | 503 | 61 | ||
AK131133.1 | - | 5 - 6 | 491 | 61 | ||
AK155816.1 | - | 5 - 6 | 507 | 61 | ||
AK170432.1 | - | 5 - 6 | 503 | 61 | ||
BC042840.1 | - | 5 - 6 | 484 | 61 | ||
BC057200.1 | - | 5 - 6 | 478 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |