Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025673.2 | NP_079949.1 | 1 - 2 | 462 | 78 | |
GenBank mRNA | AK005996.1 | - | 1 - 2 | 509 | 78 | |
AK014644.1 | - | 1 - 2 | 510 | 78 | ||
AK015543.1 | - | 1 - 2 | 506 | 78 | ||
AK028409.1 | - | 1 - 2 | 471 | 78 | ||
AK077446.1 | - | 1 - 2 | 527 | 78 | ||
AK133763.1 | - | 1 - 2 | 510 | 78 | ||
AK136194.1 | - | 1 - 2 | 87 | 78 | ||
AK167805.1 | - | 1 - 2 | 503 | 78 | ||
BC031445.1 | - | 1 - 2 | 323 | 4482612 | 78 | |
BC039909.1 | - | 1 - 2 | 460 | 78 | ||
BC051035.1 | - | 1 - 2 | 462 | 2655316 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |