Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163686.1 | NP_001157158.1 | 3 - 4 | 603 | 69 | |
NM_029761.4 | NP_084037.3 | 3 - 4 | 603 | 69 | ||
GenBank mRNA | AF418208.1 | - | 3 - 4 | 465 | 69 | |
AK012430.1 | - | 3 - 4 | 279 | 69 | ||
AK044148.1 | - | 3 - 4 | 471 | 69 | ||
BC066822.1 | - | 3 - 4 | 603 | 30021902 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |