Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK077593.1 | - | 6 - 7 | 1182 | 66 | |
AK141558.1 | - | 6 - 7 | 1213 | 66 | ||
AK165931.1 | - | 6 - 7 | 1179 | 66 | ||
BC050195.1 | - | 6 - 7 | 1158 | 30010078 | 66 | |
U70859.1 | - | 7 - 8 | 1316 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |