Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008976.2 | NP_033002.2 | 13 - 14 | 3089 | 71 | |
GenBank mRNA | AK156480.1 | - | 13 - 14 | 2835 | 71 | |
D31842.1 | - | 12 - 13 | 2686 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons |