Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009178.3 | NP_033204.2 | 7 - 8 | 609 | 59 | |
GenBank mRNA | AB061305.1 | - | 7 - 8 | 557 | 59 | |
AK008543.1 | - | 1 - 2 | 87 | 59 | ||
AK131874.1 | - | 1 - 2 | 87 | 59 | ||
BC011121.1 | - | 7 - 8 | 554 | 3497996 | 59 | |
BC050773.1 | - | 7 - 8 | 609 | 6587379 | 59 | |
D28941.1 | - | 7 - 8 | 564 | 59 | ||
X95809.1 | - | 6 - 7 | 436 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |