Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015780.2 | NP_056595.1 | 1 - 2 | 138 | 110 | |
GenBank mRNA | BC111870.1 | - | 1 - 2 | 121 | 40057075 | 110 |
M29008.1 | - | 1 - 2 | 138 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |