Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042489.2 | NP_001035954.1 | 3 - 4 | 381 | 96 | |
NM_028752.3 | NP_083028.1 | 2 - 3 | 243 | 96 | ||
GenBank mRNA | AK002854.1 | - | 2 - 3 | 244 | 96 | |
AK154880.1 | - | 3 - 4 | 273 | 96 | ||
AK155123.1 | - | 3 - 4 | 218 | 96 | ||
AK170422.1 | - | 3 - 4 | 242 | 96 | ||
BC021548.1 | - | 3 - 4 | 212 | 4913027 | 96 | |
BY703352.1 | - | 2 - 3 | 237 | 96 | ||
CF734112.1 | - | 3 - 4 | 216 | 96 | ||
CF749152.1 | - | 3 - 4 | 381 | 96 | ||
CJ164124.1 | - | 3 - 4 | 379 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |