Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133249.2 | NP_573512.1 | 5 - 6 | 1742 | 79 | |
GenBank mRNA | AF453324.1 | - | 5 - 6 | 1742 | 79 | |
AK028464.1 | - | 6 - 7 | 1792 | 79 | ||
AK086111.1 | - | 5 - 6 | 1745 | 79 | ||
BC150699.1 | - | 5 - 6 | 1722 | 9087610 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |