Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133351.3 | NP_579929.1 | 4 - 5 | 768 | 102 | |
GenBank mRNA | AB038244.1 | - | 4 - 5 | 749 | 102 | |
AF188613.1 | - | 4 - 5 | 682 | 102 | ||
AF378085.2 | - | 4 - 5 | 759 | 102 | ||
AK010640.1 | - | 4 - 5 | 651 | 102 | ||
AK078696.1 | - | 4 - 5 | 723 | 102 | ||
AY335911.1 | - | 5 - 6 | 616 | 102 | ||
BC003851.1 | - | 4 - 5 | 742 | 3600399 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |