Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133922.3 | NP_598683.3 | 10 - 11 | 1696 | 72 | |
GenBank mRNA | AB041543.1 | - | 8 - 9 | 1328 | 72 | |
AK029235.1 | - | 1 - 2 | 142 | 72 | ||
AK040525.1 | - | 10 - 11 | 1698 | 72 | ||
AK128931.1 | - | 10 - 11 | 1647 | 72 | ||
AK162934.1 | - | 10 - 11 | 1404 | 72 | ||
AK173281.1 | - | 10 - 11 | 3400 | 72 | ||
AK197252.1 | - | 1 - 2 | 44 | 72 | ||
BC066816.1 | - | 10 - 11 | 1616 | 30084911 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |