Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133950.2 | NP_598711.1 | 3 - 4 | 519 | 75 | |
GenBank mRNA | AJ278132.1 | - | 3 - 4 | 519 | 75 | |
AK013118.1 | - | 3 - 4 | 535 | 75 | ||
AK078785.1 | - | 3 - 4 | 588 | 75 | ||
AY027435.1 | - | 3 - 4 | 348 | 75 | ||
BC011370.1 | - | 3 - 4 | 431 | 3152867 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |