Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205181.1 | NP_001192110.1 | 2 - 3 | 436 | 66 | |
NM_134076.2 | NP_598837.2 | 2 - 3 | 181 | 66 | ||
GenBank mRNA | AK030250.1 | - | 2 - 3 | 377 | 66 | |
AK049366.1 | - | 2 - 3 | 162 | 66 | ||
AK152595.1 | - | 2 - 3 | 182 | 66 | ||
AK161853.1 | - | 2 - 3 | 131 | 66 | ||
AK164655.1 | - | 2 - 3 | 141 | 66 | ||
BC017532.1 | - | 2 - 3 | 116 | 4482138 | 66 | |
CN713874.1 | - | 2 - 3 | 436 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |