Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138586.3 | NP_613052.1 | 1 - 2 | 223 | 61 | |
NR_104358.1 | 1 - 2 | 223 | 61 | |||
GenBank mRNA | AK006475.1 | - | 1 - 2 | 176 | 61 | |
AK007372.1 | - | 1 - 2 | 212 | 61 | ||
AK155228.1 | - | 1 - 2 | 175 | 61 | ||
BB856072.1 | - | 1 - 2 | 225 | 61 | ||
BC034358.1 | - | 1 - 2 | 169 | 2651418 | 61 | |
BC092393.1 | - | 1 - 2 | 164 | 61 | ||
BY196593.1 | - | 1 - 2 | 221 | 61 | ||
BY198385.1 | - | 1 - 2 | 221 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |