Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083912.1 | NP_001077381.1 | 3 - 4 | 1188 | 74 | |
NM_001290542.1 | NP_001277471.1 | 3 - 4 | 1411 | 74 | ||
NM_138752.2 | NP_620091.2 | 3 - 4 | 1191 | 74 | ||
GenBank mRNA | AF465238.1 | - | 3 - 4 | 1656 | 74 | |
AK131110.1 | - | 3 - 4 | 1020 | 74 | ||
AK144152.1 | - | 3 - 4 | 1351 | 74 | ||
BC040403.1 | - | 1 - 2 | 187 | 74 | ||
BC052436.1 | - | 2 - 3 | 525 | 6837328 | 74 | |
BC056971.1 | - | 3 - 4 | 459 | 6821362 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |