Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139151.4 | NP_631890.3 | 6 - 7 | 758 | 67 | |
GenBank mRNA | AB041554.1 | - | 4 - 5 | 1337 | 67 | |
AK028080.1 | - | 6 - 7 | 727 | 67 | ||
AK043812.1 | - | 6 - 7 | 726 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |