Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144784.3 | NP_659033.1 | 8 - 9 | 892 | 64 | |
GenBank mRNA | AJ510150.1 | - | 8 - 9 | 884 | 64 | |
AK032097.1 | - | 8 - 9 | 863 | 64 | ||
AK075593.1 | - | 8 - 9 | 865 | 64 | ||
AK081715.1 | - | 8 - 9 | 892 | 64 | ||
AK146241.1 | - | 8 - 9 | 847 | 64 | ||
AK163402.1 | - | 8 - 9 | 896 | 64 | ||
AK169771.1 | - | 8 - 9 | 885 | 64 | ||
BC024429.1 | - | 8 - 9 | 895 | 64 | ||
BC024763.1 | - | 8 - 9 | 864 | 5365469 | 64 | |
BC029345.1 | - | 5 - 6 | 552 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |