Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025819.4 | NP_080095.3 | 12 - 13 | 1868 | 95 | |
GenBank mRNA | AK004803.2 | - | 12 - 13 | 1815 | 95 | |
BC055064.1 | - | 12 - 13 | 1869 | 6528686 | 95 | |
BC060204.1 | - | 12 - 13 | 1808 | 6855442 | 95 | |
CV563132.1 | - | 2 - 3 | 212 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |