Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017080.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017076.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017081.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017077.mm.1 |
TC0300002273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195499.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195499.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195500.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_001195500.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_027539.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
NM_027539.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017079.mm.1 |
TC0300002273.mm.1 |
MTA TC |