Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482246 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482198 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482200 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482202 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482203 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482248 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482199 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482201 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482247 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482197 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099930 |
BC046797.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099930 |
AY029533.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099930 |
BC043059.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099930 |
AK145762.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099930 |
AK146220.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395578 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301338 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
852708 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
247693 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47154 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647250 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69597 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
772622 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831272 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
701268 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
328020 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899124 |
AK019386.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435509 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
322982 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
362504 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380238 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341113 |
AK004955.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341113 |
AK144591.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341113 |
BC037615.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341113 |
BC048245.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341113 |
NM_178376.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
56258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547494 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
303967 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
27112 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
531557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83788 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88642 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132365 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
48374 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88253 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125582 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140441 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099930 |
BC046797.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099930 |
AY029533.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099930 |
BC043059.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099930 |
AK145762.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099930 |
AK146220.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P481546 |
BC046797.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P481546 |
AY029533.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P481546 |
BC010670.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P481546 |
BC043059.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P481546 |
AK145762.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482207 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482188 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482200 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482220 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482190 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482203 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482192 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482216 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482210 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482194 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482196 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482224 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482198 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482221 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482205 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482211 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482219 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482214 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482187 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482202 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482204 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30482193 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08479827 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08479841 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140441 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03113853 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125582 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08479806 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P481546 |
BC046797.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P481546 |
AY029533.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P481546 |
BC010670.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P481546 |
BC043059.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P481546 |
AK145762.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126956 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140441 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125582 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12289022 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12289024 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12093168 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12093166 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482246 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482197 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482199 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482200 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482248 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482247 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482198 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482203 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482202 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30482201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30481999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30481997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30482048 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482226 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482167 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482224 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11881480 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12248910 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482194 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482220 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12187139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482180 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482219 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12325239 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12369111 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12093168 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482217 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482174 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482164 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12275538 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482193 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482196 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482211 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482181 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482022 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482171 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482204 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482207 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11949464 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482198 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482179 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482178 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482221 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11881479 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482203 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482187 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482185 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482190 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12460323 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482182 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12289022 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12325240 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482186 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12179614 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482214 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30482172 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12369112 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12093163 |
- |
- |