Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286003.1 | NP_001272932.1 | 1 - 2 | 424 | 82 | |
NM_028776.5 | NP_083052.1 | 1 - 2 | 424 | 82 | ||
GenBank mRNA | AK004809.2 | - | 1 - 2 | 164 | 82 | |
AK030824.1 | - | 1 - 2 | 400 | 82 | ||
AK044100.1 | - | 1 - 2 | 385 | 82 | ||
BB653492.1 | - | 1 - 2 | 425 | 82 | ||
BC043085.1 | - | 1 - 2 | 125 | 6417440 | 82 | |
BC066800.1 | - | 1 - 2 | 109 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |