Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290148.1 | NP_001277077.1 | 10 - 11 | 981 | 97 | |
NM_028964.4 | NP_083240.2 | 17 - 18 | 2030 | 97 | ||
GenBank mRNA | AF399755.1 | - | 10 - 11 | 936 | 97 | |
AK017102.1 | - | 10 - 11 | 982 | 97 | ||
AK086865.1 | - | 5 - 6 | 563 | 97 | ||
AK162238.1 | - | 9 - 10 | 859 | 97 | ||
BC034114.1 | - | 7 - 8 | 630 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |