Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127191.2 | NP_001120663.1 | 2 - 3 | 686 | 116 | |
NM_029068.4 | NP_083344.3 | 2 - 3 | 686 | 116 | ||
GenBank mRNA | AK015872.1 | - | 2 - 3 | 647 | 116 | |
AK032055.1 | - | 2 - 3 | 643 | 116 | ||
AK166422.1 | - | 2 - 3 | 660 | 116 | ||
BC019424.1 | - | 1 - 2 | 451 | 5052234 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |