Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029352.3 | NP_083628.3 | 1 - 2 | 204 | 80 | |
GenBank mRNA | AK141667.1 | - | 1 - 2 | 227 | 80 | |
AY316312.1 | - | 1 - 2 | 204 | 80 | ||
BC100309.1 | - | 1 - 2 | 204 | 30787409 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |