Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144547.2 | NP_653130.2 | 1 - 2 | 158 | 75 | |
GenBank mRNA | AF503863.1 | - | 1 - 2 | 105 | 75 | |
BB868995.1 | - | 1 - 2 | 160 | 75 | ||
BC119571.1 | - | 1 - 2 | 89 | 40057985 | 75 | |
BC119572.2 | - | 10 - 11 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |