Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300014062.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014062.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014062.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014071.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014071.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014071.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014071.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014060.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014060.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014060.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014060.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014061.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014061.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014061.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014063.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014063.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014063.mm.1 |
TC1300002184.mm.1 |
MTA TC |