Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277863.1 | NP_001264792.1 | 9 - 10 | 990 | 91 | |
NM_010021.5 | NP_034151.3 | 10 - 11 | 1041 | 91 | ||
GenBank mRNA | AK014854.1 | - | 10 - 11 | 1021 | 91 | |
AK033183.1 | - | 10 - 11 | 1021 | 91 | ||
AK160855.1 | - | 10 - 11 | 900 | 91 | ||
U38690.1 | - | 10 - 11 | 1156 | 91 | ||
U46694.1 | - | 10 - 11 | 937 | 91 | ||
X95724.1 | - | 10 - 11 | 930 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |