Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285947.1 | NP_001272876.1 | 5 - 6 | 931 | 72 | |
NM_001285948.1 | NP_001272877.1 | 6 - 7 | 1081 | 72 | ||
NM_018887.4 | NP_061375.1 | 6 - 7 | 1081 | 72 | ||
GenBank mRNA | AF237981.1 | - | 6 - 7 | 930 | 72 | |
AK031733.1 | - | 6 - 7 | 1029 | 72 | ||
BC021418.1 | - | 6 - 7 | 952 | 5025117 | 72 | |
BC037663.1 | - | 5 - 6 | 780 | 5352740 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |