Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021284.6 | NP_067259.4 | 1 - 2 | 183 | 104 | |
GenBank mRNA | AK155064.1 | - | 1 - 2 | 184 | 104 | |
AK164065.1 | - | 1 - 2 | 184 | 104 | ||
BC004642.1 | - | 1 - 2 | 171 | 3158212 | 104 | |
BC010202.1 | - | 1 - 2 | 171 | 3158212 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |