Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199020.1 | NP_001185949.1 | 4 - 5 | 704 | 107 | |
NM_021421.4 | NP_067396.3 | 3 - 4 | 839 | 107 | ||
NR_037578.1 | 3 - 4 | 798 | 107 | |||
NR_037579.1 | 2 - 3 | 513 | 107 | |||
GenBank mRNA | AB041602.1 | - | 2 - 3 | 499 | 107 | |
AK077039.1 | - | 3 - 4 | 785 | 107 | ||
AK088834.1 | - | 2 - 3 | 309 | 107 | ||
AK135709.1 | - | 4 - 5 | 701 | 107 | ||
AK158731.1 | - | 3 - 4 | 855 | 107 | ||
AK166619.1 | - | 4 - 5 | 703 | 107 | ||
AK169653.1 | - | 2 - 3 | 423 | 107 | ||
AK205192.1 | - | 1 - 2 | 71 | 107 | ||
AK206765.1 | - | 1 - 2 | 87 | 107 | ||
BC024907.1 | - | 3 - 4 | 799 | 2812124 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |